Merck
  • Enhanced sensitivity to drug-induced QT interval lengthening in patients with heart failure due to left ventricular systolic dysfunction.
Enhanced sensitivity to drug-induced QT interval lengthening in patients with heart failure due to left ventricular systolic dysfunction.
Journal of clinical pharmacology (2011-11-03)
James E Tisdale, Brian R Overholser, Heather A Wroblewski, Kevin M Sowinski, Kwadwo Amankwa, Steven Borzak, Joanna R Kingery, Rita Coram, Douglas P Zipes, David A Flockhart, Richard J Kovacs
ABSTRACT
Patients with heart failure (HF) are at increased risk for drug-induced torsades de pointes (TdP) due to unknown mechanisms. Our objective was to determine if sensitivity to drug-induced QT interval lengthening is enhanced in patients with HF. In this multicenter, prospective study, 15 patients with atrial fibrillation or flutter requiring conversion to sinus rhythm were enrolled: 6 patients with New York Heart Association class II to III HF (mean ejection fraction [EF], 30% ± 9%), and 9 controls (mean EF, 53% ± 6%). Patients received ibutilide 1 mg intravenously. Blood samples and 12-lead electrocardiograms were obtained prior to and during 48 hours postinfusion. Serum ibutilide concentrations at 50% maximum effect on Fridericia-corrected QT (QT(F)) intervals (EC(50)) were determined, and areas under the effect (QT(F) interval vs time) curves (AUECs) were calculated. Ibutilide concentration-QT(F) relationships were best described by a sigmoidal E(max) model with a hypothetical effect compartment. Median [interquartile range] AUEC from 0 to 4 hours was larger in the HF group than in controls (1.86 [1.86-1.93] vs 1.82 [1.81-1.84] s·h; P = .04). Median EC(50) was lower in the HF group (0.48 [0.46-0.49] vs 1.85 [1.10-3.23] μg/L; P = .008). Sensitivity to drug-induced QT interval lengthening is enhanced in patients with systolic HF, which may contribute to the increased risk of drug-induced TdP.
Sigma-Aldrich
Ibutilide hemifumarate salt, ≥98% (HPLC)