- Antiproliferative activity of N(6)-isopentenyladenosine on HCT-15 colon carcinoma cell line.
Antiproliferative activity of N(6)-isopentenyladenosine on HCT-15 colon carcinoma cell line.
Nucleic acid therapeutics (2011-10-19)
Mehdi Rajabi, Jamshid Mehrzad, Elena Gorincioi, Enzo Santaniello
PMID22004416
ABSTRACT
N(6)-Isopentenyladenosine (iPA), a member of the cytokinin family of plant hormones, exerts remarkable inhibition on tumor cell proliferation and apoptosis in several tumor cell lines. In this study, we report that iPA is able to inhibit the proliferation and promotes apoptosis in HCT-15 human colon cancer cells in a dose-dependent manner with a concentration of 2.5 μM, which causes 50% inhibition of cell viability. The cell cycle analysis by flow cytometry showed that iPA-induced growth arrest could be associated to apoptosis. Moreover, suppression of clonogenic activity occurs after exposure to iPA at a concentration of 2.5 μM for HCT-15.