Merck
  • Simultaneous measurement of plasma vitamin D(3) metabolites, including 4β,25-dihydroxyvitamin D(3), using liquid chromatography-tandem mass spectrometry.
Simultaneous measurement of plasma vitamin D(3) metabolites, including 4β,25-dihydroxyvitamin D(3), using liquid chromatography-tandem mass spectrometry.
Analytical biochemistry (2011-07-26)
Zhican Wang, Tauri Senn, Tom Kalhorn, Xi Emily Zheng, Songmao Zheng, Connie L Davis, Mary F Hebert, Yvonne S Lin, Kenneth E Thummel
ABSTRACT
Simultaneous and accurate measurement of circulating vitamin D metabolites is critical to studies of the metabolic regulation of vitamin D and its impact on health and disease. To that end, we have developed a specific liquid chromatography-tandem mass spectrometry (LC-MS/MS) method that permits the quantification of major circulating vitamin D(3) metabolites in human plasma. Plasma samples were subjected to a protein precipitation, liquid-liquid extraction, and Diels-Alder derivatization procedure prior to LC-MS/MS analysis. Importantly, in all human plasma samples tested, we identified a significant dihydroxyvitamin D(3) peak that could potentially interfere with the determination of 1α,25-dihydroxyvitamin D(3) [1α,25(OH)(2)D(3)] concentrations. This interfering metabolite has been identified as 4β,25-dihydroxyvitamin D(3) [4β,25(OH)(2)D(3)] and was found at concentrations comparable to 1α,25(OH)(2)D(3). Quantification of 1α,25(OH)(2)D(3) in plasma required complete chromatographic separation of 1α,25(OH)(2)D(3) from 4β,25(OH)(2)D(3). An assay incorporating this feature was used to simultaneously determine the plasma concentrations of 25OHD(3), 24R,25(OH)(2)D(3), 1α,25(OH)(2)D(3), and 4β,25(OH)(2)D(3) in healthy individuals. The LC-MS/MS method developed and described here could result in considerable improvement in quantifying 1α,25(OH)(2)D(3) as well as monitoring the newly identified circulating metabolite, 4β,25(OH)(2)D(3).
Sigma-Aldrich
(24R)-24,25-Dihydroxyvitamin D3, ≥98% (vitamin + pre-vitamin, HPLC)