Merck
  • Systematic evaluation of new chiral stationary phases for supercritical fluid chromatography using a standard racemate library.
Systematic evaluation of new chiral stationary phases for supercritical fluid chromatography using a standard racemate library.
Journal of chromatography. A (2009-10-24)
Zainab Pirzada, Michelle Personick, Mirlinda Biba, Xiaoyi Gong, Lili Zhou, Wes Schafer, Christian Roussel, Christopher J Welch
ABSTRACT
A systematic approach to the evaluation of new chiral stationary phases (CSPs) for supercritical fluid chromatography (SFC) using a standard library of racemic analytes is described. A standard library of racemic analytes representing a variety of functional group classes was assembled from a mixture of proprietary and commercial compounds. The library is dispensed and stored in a convenient 96-well microplate format to facilitate ease of use, and to minimize the amount of analyte required for analysis. Automated SFC screening was performed on both established CSPs in common use, as well as a group of six recently commercialized CSPs. Screening results were archived in a structure-searchable database that allows convenient comparison of performance data to determine which CSPs shows the best performance.