Merck
  • A new member of the alkaline phosphatase superfamily with a formylglycine nucleophile: structural and kinetic characterisation of a phosphonate monoester hydrolase/phosphodiesterase from Rhizobium leguminosarum.
A new member of the alkaline phosphatase superfamily with a formylglycine nucleophile: structural and kinetic characterisation of a phosphonate monoester hydrolase/phosphodiesterase from Rhizobium leguminosarum.
Journal of molecular biology (2008-09-17)
Stefanie Jonas, Bert van Loo, Marko Hyvönen, Florian Hollfelder
ABSTRACT
The alkaline phosphatase superfamily comprises a large number of hydrolytic metalloenzymes such as phosphatases and sulfatases. We have characterised a new member of this superfamily, a phosphonate monoester hydrolase/phosphodiesterase from Rhizobium leguminosarum (R/PMH) both structurally and kinetically. The 1.42 A crystal structure shows structural homology to arylsulfatases with conservation of the core alpha/beta-fold, the mononuclear active site and most of the active-site residues. Sulfatases use a unique formylglycine nucleophile, formed by posttranslational modification of a cysteine/serine embedded in a signature sequence (C/S)XPXR. We provide mass spectrometric and mutational evidence that R/PMH is the first non-sulfatase enzyme shown to use a formylglycine as the catalytic nucleophile. R/PMH hydrolyses phosphonate monoesters and phosphate diesters with similar efficiency. Burst kinetics suggest that substrate hydrolysis proceeds via a double-displacement mechanism. Kinetic characterisation of active-site mutations establishes the catalytic contributions of individual residues. A mechanism for substrate hydrolysis is proposed on the basis of the kinetic data and structural comparisons with E. coli alkaline phosphatase and Pseudomonas aeruginosa arylsulfatase. R/PMH represents a further example of conservation of the overall structure and mechanism within the alkaline phosphatase superfamily.