Merck
  • Effects of in utero and lactational exposure to triphenyltin chloride on pregnancy outcome and postnatal development in rat offspring.
Effects of in utero and lactational exposure to triphenyltin chloride on pregnancy outcome and postnatal development in rat offspring.
Toxicology (2007-07-24)
Konstanze Grote, Carolin Hobler, Anderson J M Andrade, Simone Wichert Grande, Christine Gericke, Chris E Talsness, Klaus E Appel, Ibrahim Chahoud
ABSTRACT
The organotin compound (OTC) triphenyltin (TPT) is used extensively as a herbicide, pesticide and fungicide in agriculture as well as, together with tributyltin (TBT), in marine antifouling paints. We studied the effects of in utero exposure to 2 or 6 mg triphenyltinchloride (TPTCl)/kgb.w. on pregnancy outcome and postnatal development in rat offspring. Gravid Wistar rats were treated per gavage from gestational day 6 until the end of lactation. In the 6 mg TPTCl dose group gestational mortality in dams as well as an increased incidence of anticipated and delayed parturition was observed. Furthermore, treatment resulted in a significant increase in perinatal mortality, a decrease in lactational body weight gain as well as in delayed physical maturation of offspring. Similarily, exposure to 2mg TPTCl/kgb.w. resulted in a significant increase in perinatal mortality and in delayed eye opening. Lactational body weight gain and other landmarks of physical maturation were unaffected in the low dose group. We conclude, that in utero exposure to TPTCl at the described dose levels severely affected pregnancy outcome and perinatal survival of offspring. These results were unexpected, as in two earlier studies with pubertal rats TPTCl at the same dose levels no signs of general toxicity were observed.