Merck
  • [Determination of anions in the products of ozonation of reactive dye by ion chromatography].
[Determination of anions in the products of ozonation of reactive dye by ion chromatography].
Se pu = Chinese journal of chromatography (2005-12-20)
Fei-Fang Zhang, Xin-Miao Liang, Qing Zhang, Ji-Ping Chen, Ayfer Yediler, Antonius Kettrup
ABSTRACT
A method for the determination of anions in the degradation products of C. I. Reactive Red 120 by ozonation was investigated. The sample can be pretreated with a Dionex OnGuard P column, which has high selectivity for removing phenols, azo group-contaning compounds, aromatic carboxylic aicds etc. For the separation of organic and inorganic anions in the products, the anion exchange chromatograhy with gradient elution of NaOH was used. All the species were detected by both suppressed conductivity and UV absorbance detectors. The sulfate, oxalate, chloride, nitrate and formate could be identified even in very low concentration with ion chromatography, within 18 min, with the recoveries between 91.6% - 108.3%. The results indicate that the method is reliable, simple, rapid and especially sensitive. In combining with the determination results of conventional parameters of the solution of Reactive Red 120, a preliminary elucidation of the degradation mechanism
Sigma-Aldrich
Reactive Red 120