Merck
  • Clozapine-induced salivation: interaction with N-desmethylclozapine and amisulpride in an experimental rat model.
Clozapine-induced salivation: interaction with N-desmethylclozapine and amisulpride in an experimental rat model.
European journal of oral sciences (2011-07-06)
Tania Godoy, Alessandro Riva, Jörgen Ekström
ABSTRACT
Many drugs (e.g. amisulpride) have been used to treat troublesome clozapine-induced salivation; however, varying success has been achieved in this respect, probably because, until recently, the salivatory action of clozapine has been largely unexplained. In the rat, clozapine and its main metabolite, N-desmethylclozapine, were found to exert mixed secretory actions: excitatory, through muscarinic acetylcholine M1-receptors giving rise to a long-lasting, low-level flow of saliva; and inhibitory, through muscarinic M3-receptors and α(1) -adrenoceptors reducing the parasympathetically and sympathetically nerve-evoked flow of saliva. The aim of the present study was to define the interactions between clozapine and N-desmethylclozapine, and clozapine and amisulpride, with respect to the excitatory response. Submandibular glands, sensitized by chronic parasympathetic preganglionic denervation, were studied in pentobarbitone-anaesthetized rats. To prevent clozapine from being metabolized to N-desmethylclozapine by hepatic enzymes, the liver was, under terminal anaesthesia, excluded from the circulation. The weak receptor-stimulating clozapine prevented the strong receptor-stimulating N-desmethylclozapine, at specific ratios in humans and in rats, from exerting its full agonistic action. In conclusion, the contribution of N-desmethylclozapine to the clozapine-induced sialorrhoea was, at most, only partly additive. Furthermore, the present experimental set-up failed to demonstrate any anti-salivatory action of amisulpride on the clozapine-induced flow of saliva.
Product Description
Supelco
N-Desmethylclozapine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
N-Desmethylclozapine