Merck
Improved method for the synthesis of 1- or 3-acyl-sn-glycerols.
Journal of lipid research (1987-04-01)
D R Kodali
ABSTRACT
Optically active 1- or 3-acyl-sn-glycerols were synthesized from 2,3- or 1,2-isopropylidene-sn-glycerols, respectively. The 2,3- or 1,2-isopropylidene-sn-glycerols were condensed with appropriate long saturated or unsaturated fatty acids and the resulting acyl isopropylidene compounds were treated with dimethylboronbromide at - 50 degrees C to give the title compounds. The ketal cleavage of acyl isopropylidene-sn-glycerols by dimethylboronbromide to produce the long 1- or 3-acyl-sn-glycerols was effective and gave good yields (70-90%). The reaction conditions were mild and there was no acyl migration, as shown by optical rotation of the monoacyl-sn-glycerols. The synthesis of 2,3-isopropylidene-sn-glycerol was improved to give an overall yield of 40% from L-arabinose. L-Arabinose was first converted to its 1,1'-diethylmercapto derivative and then condensed with 2-methoxypropene to yield 1,1'-diethyl-mercapto-4,5-isopropylidene-L-arabinose. Oxidation of this compound with sodium periodate followed by reduction with sodium borohydride under alkaline conditions yielded 2,3-isopropylidene-sn-glycerol [alpha]22D = -14.90 degrees, neat (Lit. 8 [alpha]22D = -14.5 degrees, neat; 14 [alpha]25D = -10.8 degrees; methanol C, 16.9). The optical purity of isopropylidene-sn-glycerols was determined as benzoyl derivatives on a high performance liquid chromatographic column packed with a chiral stationary phase.
Sigma-Aldrich
DL-1,2-Isopropylideneglycerol, ≥97.0%
Sigma-Aldrich
(R)-(−)-2,2-Dimethyl-1,3-dioxolane-4-methanol, 98%
Sigma-Aldrich
(S)-(+)-1,2-Isopropylideneglycerol, 98%, optical purity ee: 99% (GLC)