- The use of indentometry to study the effect of agents known to increase skin c-AMP content.
The use of indentometry to study the effect of agents known to increase skin c-AMP content.
Experientia (1985-03-15)
A Hartzshtark, S Dikstein
PMID2982648
ABSTRACT
Local, externally applied pharmacological agents which are assumed to raise the c-AMP level, decrease the low pressure indentation value of the forehead skin of certain human volunteers.