- [Protection of 2'-hydroxyls in the chemical synthesis of oligoribonucleotides].
[Protection of 2'-hydroxyls in the chemical synthesis of oligoribonucleotides].
Bioorganicheskaia khimiia (2013-07-13)
A V Aralov, O G Chakhmakhchieva
PMID23844504
ABSTRACT
The review is devoted to the chemical synthesis of oligoribonucleotides and the protecting groups used. In particular the existent methods of blocking 2'-OH function in nucleotide monomers for the RNA synthesis are discussed in detail.