Merck
  • Phytoremediation of hexachlorocyclohexane (HCH)-contaminated soils using Cytisus striatus and bacterial inoculants in soils with distinct organic matter content.
Phytoremediation of hexachlorocyclohexane (HCH)-contaminated soils using Cytisus striatus and bacterial inoculants in soils with distinct organic matter content.
Environmental pollution (Barking, Essex : 1987) (2013-04-16)
Cristina Becerra-Castro, Petra S Kidd, Beatriz Rodríguez-Garrido, Carmela Monterroso, Paula Santos-Ucha, Angeles Prieto-Fernández
ABSTRACT
The performance of Cytisus striatus in association with different microbial inoculant treatments on the dissipation of the insecticide hexachlorocyclohexane (HCH) was studied. Two soils with different organic matter (A and B soil) content were spiked with 0 or 65 mg HCH kg(-1). Plants were either not inoculated (NI), or inoculated with the endophyte Rhodococcus erythropolis ET54b and the HCH-degrader Sphingomonas sp. D4 separately or in combination (ET, D4 and ETD4). Unplanted pots were also established. HCH phytotoxicity was more pronounced in the B soil. Soil HCH concentrations in unplanted pots were similar to initial concentrations, whereas concentrations were reduced after plant growth: by 20% and 8% in A and B soil, respectively. Microbial inoculants also modified HCH dissipation, although effects were soil-dependent. Inoculation with the combination of strains (ETD4) led to a significant enhancement in HCH dissipation: up to 53% in the A soil and 43% in the B soil.
Product Description
Supelco
Lindane, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Lindane, PESTANAL®, analytical standard
Supelco
γ-BHC, certified reference material, 1000 μg/mL in methanol