Merck
  • Randomized controlled trial of cetuximab plus chemotherapy for patients with KRAS wild-type unresectable colorectal liver-limited metastases.
Randomized controlled trial of cetuximab plus chemotherapy for patients with KRAS wild-type unresectable colorectal liver-limited metastases.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology (2013-04-10)
Le-Chi Ye, Tian-Shu Liu, Li Ren, Ye Wei, De-Xiang Zhu, Sheng-Yong Zai, Qing-Hai Ye, Yiyi Yu, Bo Xu, Xin-Yu Qin, Jianmin Xu
ABSTRACT
To assess the effects of cetuximab plus chemotherapy as first-line treatment for unresectable colorectal liver metastases (CLMs). After resection of their primary tumors, patients with KRAS wild-type synchronous nonresectable liver-limited metastases from colorectal cancer were randomly assigned to receive chemotherapy (FOLFIRI [fluorouracil, leucovorin, and irinotecan] or mFOLFOX6 [modified fluorouracil, leucovorin, and oxaliplatin]) plus cetuximab (arm A) or chemotherapy alone (arm B). The primary end point was the rate of patients converted to resection for liver metastases. Secondary end points included tumor response and survival. The intent-to-treat population comprised 138 patients; 70 patients were randomly assigned to arm A and 68 to arm B. After a median of 25.0 months of follow-up, the 3-year overall survival (OS) rate and median survival time (MST) for all patients were 30% and 24.4 months, respectively. The R0 resection rates for liver metastases were 25.7% (18 of 70 patients) in arm A and 7.4% (five of 68 patients) in arm B, which were significantly different (P < .01). Patients in arm A had improved objective response rates (57.1% v 29.4%; P < .01), increased 3-year OS rate (41% v 18%; P = .013) and prolonged MST (30.9 v 21.0 months; P = .013) compared with those in arm B. In addition, in arm A, patients who had resection of liver metastases had a significantly improved MST (46.4 v 25.7 months; P < .01) compared with those who did not undergo surgery. For patients with initially unresectable KRAS wild-type CLMs, cetuximab combined with chemotherapy improved the resectability of liver metastases and improved response rates and survival compared with chemotherapy alone.
Sigma-Aldrich
Folinic acid calcium salt hydrate, BioXtra, ≥99.0% (HPLC)