Merck
  • Squaramide-tertiary amine catalyzed asymmetric cascade sulfa-Michael/Michael addition via dynamic kinetic resolution: access to highly functionalized chromans with three contiguous stereocenters.
Squaramide-tertiary amine catalyzed asymmetric cascade sulfa-Michael/Michael addition via dynamic kinetic resolution: access to highly functionalized chromans with three contiguous stereocenters.
Organic letters (2013-03-08)
Wen Yang, Yi Yang, Da-Ming Du
ABSTRACT
An efficient asymmetric cascade sulfa-Michael/Michael addition reaction catalyzed by a chiral bifunctional squaramide-tertiary amine catalyst has been developed. This organocatalytic cascade reaction provides easy access to highly functionalized chromans with three contiguous stereocenters, including one quaternary center. In addition, a novel cascade sulfa Michael/retro-sulfa-Michael/sulfa-Michael/Michael reaction process, involving dynamic kinetic resolution, is described.
Sigma-Aldrich
3,4-Dihydroxy-3-cyclobutene-1,2-dione, ≥99.0% (HPLC)