Merck
  • Total syntheses of multiple cladiellin natural products by use of a completely general strategy.
Total syntheses of multiple cladiellin natural products by use of a completely general strategy.
The Journal of organic chemistry (2012-12-12)
J Stephen Clark, Raphaëlle Berger, Stewart T Hayes, Hans Martin Senn, Louis J Farrugia, Lynne H Thomas, Angus J Morrison, Luca Gobbi
ABSTRACT
The enantioselective total syntheses of 10 cladiellin natural products have been completed, starting from the known allylic alcohol (+)-14, which can be prepared in large quantities. The bridged tricyclic core of the cladiellins has been constructed via three ring-forming reactions: (i) an intramolecular reductive cyclization between an aldehyde and an unsaturated ester, mediated by samarium(II) iodide, to form a tetrahydropyranol; (ii) reaction of a metal carbenoid, generated from a diazo ketone, with an ether to produce an ylide-like intermediate that rearranges to produce E- or Z-oxabicyclo[6.2.1]-5-undecen-9-one; and (iii) a Diels-Alder cycloaddition reaction to construct the third ring found in the core structure of the cladiellins. The key ring-forming reaction, in which a diazo ketone is converted into a bridged bicyclic ether, can be tuned to give either of the isomeric oxabicyclo[6.2.1]-5-undecen-9-ones as the major product by switching from a copper to a rhodium catalyst and selecting the appropriate reaction conditions. The tricyclic products obtained from the three-step sequence involving the Diels-Alder cycloaddition reaction can be employed as advanced intermediates to prepare a wide range of cladiellin natural products.
Sigma-Aldrich
Allyl alcohol, ≥98.5%
Sigma-Aldrich
Allyl alcohol, ≥99%