Merck
  • Auxin reflux between the endodermis and pericycle promotes lateral root initiation.
Auxin reflux between the endodermis and pericycle promotes lateral root initiation.
The EMBO journal (2012-11-28)
Peter Marhavý, Marleen Vanstraelen, Bert De Rybel, Ding Zhaojun, Malcolm J Bennett, Tom Beeckman, Eva Benková
ABSTRACT
Lateral root (LR) formation is initiated when pericycle cells accumulate auxin, thereby acquiring founder cell (FC) status and triggering asymmetric cell divisions, giving rise to a new primordium. How this auxin maximum in pericycle cells builds up and remains focused is not understood. We report that the endodermis plays an active role in the regulation of auxin accumulation and is instructive for FCs to progress during the LR initiation (LRI) phase. We describe the functional importance of a PIN3 (PIN-formed) auxin efflux carrier-dependent hormone reflux pathway between overlaying endodermal and pericycle FCs. Disrupting this reflux pathway causes dramatic defects in the progress of FCs towards the next initiation phase. Our data identify an unexpected regulatory function for the endodermis in LRI as part of the fine-tuning mechanism that appears to act as a check point in LR organogenesis after FCs are specified.
Product Description
Supelco
1-Naphthaleneacetic acid, PESTANAL®, analytical standard
Sigma-Aldrich
1-Naphthaleneacetic acid, suitable for plant cell culture, BioReagent, ≥95%, crystalline
Sigma-Aldrich
1-Naphthaleneacetic acid, technical grade
Sigma-Aldrich
1-Naphthylacetic acid, BioReagent, 1 mg/mL, suitable for plant cell culture