Merck
  • CO2 superabsorption in a paddlewheel-type Ru dimer chain compound: gate-open performance dependent on inter-chain interactions.
CO2 superabsorption in a paddlewheel-type Ru dimer chain compound: gate-open performance dependent on inter-chain interactions.
Chemical communications (Cambridge, England) (2012-11-24)
Wataru Kosaka, Kayo Yamagishi, Hiroki Yoshida, Ryotaro Matsuda, Susumu Kitagawa, Masaki Takata, Hitoshi Miyasaka
ABSTRACT
A porous one-dimensional coordination chain compound, [Ru(2)(p-F-PhCO(2))(4)(phz)] (; p-F-PhCO(2)(-) = para-fluorobenzoate; phz = phenazine) derived by drying its nitrobenzene-solvated compound, specifically adsorbs CO(2) at 195 K in a stepwise sorption manner.
Product Description
Supelco
Nitrobenzene, PESTANAL®, analytical standard
Sigma-Aldrich
Nitrobenzene, ACS reagent, ≥99.0%
Sigma-Aldrich
Nitrobenzene, ReagentPlus®, 99%