Merck
  • Effect of 20(S)-ginsenoside Rg3 on streptozotocin-induced experimental type 2 diabetic rats: a urinary metabonomics study by rapid-resolution liquid chromatography/mass spectrometry.
Effect of 20(S)-ginsenoside Rg3 on streptozotocin-induced experimental type 2 diabetic rats: a urinary metabonomics study by rapid-resolution liquid chromatography/mass spectrometry.
Rapid communications in mass spectrometry : RCM (2012-11-06)
Jun Niu, Zi-Feng Pi, Hao Yue, Hongmei Yang, Yang Wang, Qing Yu, Shu-Ying Liu
ABSTRACT
20(S)-ginsenoside Rg3 is an active component of Panax ginseng. It is known that 20(S)-ginsenoside Rg3 has a protective effect against hyperglycemia, obesity and diabetes in vivo, but the precise mechanisms of these actions have not yet been entirely elucidated. A urinary metabonomics method based on rapid-resolution liquid chromatography/mass spectrometry (RRLC/MS) was developed to investigate the effect of 20(S)-ginsenoside Rg3 on type 2 diabetic rats. With multivariate statistical analysis, a clear separation between type 2 diabetic rats and those treated with 20(S)-ginsenoside Rg3 was achieved. Six potential biomarkers were found and identified. This work shows that the mechanism of the effect of 20(S)-ginsenoside Rg3 on type 2 diabetes may be involved with the regulation of nucleic acid metabolism, energy metabolism and gut flora metabolism.
Sigma-Aldrich
Ginsenoside Rg3, ≥98% (HPLC)
Supelco
Ginsenoside Rg3, analytical standard