Merck
  • A comparison of dilute aqueous p-toluenesulfonic and sulfuric acid pretreatments and saccharification of corn stover at moderate temperatures and pressures.
A comparison of dilute aqueous p-toluenesulfonic and sulfuric acid pretreatments and saccharification of corn stover at moderate temperatures and pressures.
Bioresource technology (2012-10-03)
Ananda S Amarasekara, Bernard Wiredu
ABSTRACT
Single step pretreatment-saccharification of corn stover was investigated in aqueous p-toluenesulfonic and sulfuric acid media. Dilute aqueous solution of p-toluenesulfonic acid was a better catalyst than aqueous sulfuric acid of the same H(+) ion concentration for single step pretreatment-saccharification of corn stover at moderate temperatures and pressures. For example, 100mg corn stover heated at 150°C for 1h in 0.100 M H(+) aqueous sulfuric acid produced 64 μmol of total reducing sugars (TRS), whereas the sample heated in 0.100 M H(+)p-toluenesulfonic acid produced 165 μmol of TRS under identical conditions. Glucose yield showed a similar trend, as aq. sulfuric acid and p-toluene sulfonic acid media produced 29 and 35 μmol of glucose respectively after 2.5h. Higher catalytic activity of p-toluenesulfonic acid may be due to an interaction with biomass, supported by repulsion of hydrophobic tolyl group by the aqueous phase.
Sigma-Aldrich
Sodium p-toluenesulfinate, 95%
Sigma-Aldrich
Sodium p-toluenesulfinate, purum, anhydrous, ≥96.0% (NT)