Merck
  • Synthesis and biological evaluation of novel piperazine derivatives of flavone as potent anti-inflammatory and antimicrobial agent.
Synthesis and biological evaluation of novel piperazine derivatives of flavone as potent anti-inflammatory and antimicrobial agent.
Bioorganic & medicinal chemistry letters (2012-09-18)
Girish D Hatnapure, Ashish P Keche, Atish H Rodge, Satish S Birajdar, Rajesh H Tale, Vandana M Kamble
ABSTRACT
A series of novel 6-methoxy-2-(piperazin-1-yl)-4H-chromen-4-one and 5,7-dimethoxy-2-(piperazin-1-ylmethyl)-4H-chromen-4-one derivatives of biological interest were prepared and screened for their pro-inflammatory cytokines (TNF-α and IL-6) and antimicrobial activity (antibacterial and antifungal). Among all the compound screened (5a-j and 10k-t), the compounds 5c, 5g, 5h, 10l, 10m, 10n and 10r found to have promising anti-inflammatory activity (up to 65-87% TNF-α and 70-93% IL-6 inhibitory activity) at concentration of 10 μM with reference to standard dexamethasone (71% TNF-a and 84% IL-6 inhibitory activities at 1 μM) while the compounds 5b, 5i, 5j, 10s and 10t found to be potent antimicrobial agent showing even 2 to 2.5-fold more potency than that of standard ciprofloxacin and miconazole at the same MIC value of 10 μg/mL.
Product Description
Supelco
Piperazine hexahydrate, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Piperazine, BioUltra, anhydrous, ≥99.0% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Piperazine, ReagentPlus®, 99%
Pricing and availability is not currently available.