Merck
  • Biochemical and molecular characterization of the gentisate transporter GenK in Corynebacterium glutamicum.
Biochemical and molecular characterization of the gentisate transporter GenK in Corynebacterium glutamicum.
PloS one (2012-07-19)
Ying Xu, Song-He Wang, Hong-Jun Chao, Shuang-Jiang Liu, Ning-Yi Zhou
ABSTRACT
Gentisate (2,5-dihydroxybenzoate) is a key ring-cleavage substrate involved in various aromatic compounds degradation. Corynebacterium glutamicum ATCC13032 is capable of growing on gentisate and genK was proposed to encode a transporter involved in this utilization by its disruption in the restriction-deficient mutant RES167. Its biochemical characterization by uptake assay using [(14)C]-labeled gentisate has not been previously reported. In this study, biochemical characterization of GenK by uptake assays with [(14)C]-labeled substrates demonstrated that it specifically transported gentisate into the cells with V(max) and K(m) of 3.06 ± 0.16 nmol/min/mg of dry weight and 10.71 ± 0.11 µM respectively, and no activity was detected for either benzoate or 3-hydoxybenzoate. When GenK was absent in strain RES167 ΔgenK, it retained 85% of its original transport activity at pH 6.5 compared to that of strain RES167. However, it lost 79% and 88% activity at pH 7.5 and 8.0, respectively. A number of competing substrates, including 3-hydroxybenzoate, benzoate, protocatechuate and catechol, significantly inhibited gentisate uptake by more than 40%. Through site-directed mutagenesis, eight amino acid residues of GenK, Asp-54, Asp-57 and Arg-386 in the hydrophobic transmembrane regions and Arg-103, Trp-309, Asp-312, Arg-313 and Ile-317 in the hydrophilic cytoplasmic loops were shown to be important for gentisate transport. When conserved residues Asp-54 and Asp-57 respectively were changed to glutamate, both mutants retained approximately 50% activity and were able to partially complement the ability of strain RES167 ΔgenK to grow on gentisate. Our results demonstrate that GenK is an active gentisate transporter in Corynebacterium glutamicum ATCC13032. The GenK-mediated gentisate transport was also shown to be a limiting step for the gentisate utilization by this strain. This enhances our understanding of gentisate transport in the microbial degradation of aromatic compounds.
Sigma-Aldrich
2,5-Dihydroxybenzoic acid, 98%
Supelco
2,5-Dihydroxybenzoic acid, >99.0% (HPLC)
Supelco
2,5-Dihydroxybenzoic acid, ≥99.5% (HPLC), Ultra pure
Sigma-Aldrich
Gentisic acid sodium salt hydrate, ≥98%