Merck
  • Polyelectrolyte-multilayer-supported Au@Ag core-shell nanoparticles with high catalytic activity.
Polyelectrolyte-multilayer-supported Au@Ag core-shell nanoparticles with high catalytic activity.
Advanced materials (Deerfield Beach, Fla.) (2012-07-19)
Xin Zhang, Zhaohui Su
ABSTRACT
Au-Ag bimetallic core-shell nanoparticles are synthesized in situ in polyelectrolyte multilayers via a sequential ion-exchange/reduction process. The Au-Ag bimetallic nanoparticles show a higher catalytic activity than Au and Ag monometallic ones for the reduction of 4-nitrophenol by NaBH(4) .
Sigma-Aldrich
Poly(diallyldimethylammonium chloride) solution, average Mw 200,000-350,000 (medium molecular weight), 20 wt. % in H2O
Sigma-Aldrich
Poly(diallyldimethylammonium chloride) solution, average Mw 400,000-500,000 (high molecular weight), 20 wt. % in H2O
Sigma-Aldrich
Poly(diallyldimethylammonium chloride) solution, 20 wt. % in H2O
Sigma-Aldrich
Poly(diallyldimethylammonium chloride) solution, average Mw <100,000 (very low molecular weight), 35 wt. % in H2O
Sigma-Aldrich
Poly(sodium 4-styrenesulfonate) solution, average Mw ~200,000, 30 wt. % in H2O
Sigma-Aldrich
Poly(sodium 4-styrenesulfonate), average Mw ~1,000,000, powder
Sigma-Aldrich
Poly(sodium 4-styrenesulfonate), average Mw ~70,000
Sigma-Aldrich
Poly(sodium 4-styrenesulfonate) solution, average Mw ~1,000,000, 25 wt. % in H2O
Sigma-Aldrich
Poly(sodium 4-styrenesulfonate) solution, average Mw ~70,000, 30 wt. % in H2O