- Polymer micelle-directed growth of BaCO3 spiral nanobelts.
Polymer micelle-directed growth of BaCO3 spiral nanobelts.
Chemical communications (Cambridge, England) (2012-07-19)
Wenjie Zhu, Chunhua Cai, Jiaping Lin, Liquan Wang, Lili Chen, Zeliang Zhuang
PMID22806094
ABSTRACT
We discovered that micelles of a thermo-responsive polypeptide-based copolymer are able to direct growth of barium carbonate (BaCO(3)) in the form of nanobelts. The BaCO(3) nanobelts tend to grow around a formed crystal, and curl into a spiral superstructure.