Merck
  • Simultaneous determination of methocarbamol and ibuprofen or diclofenac potassium using mean centering of the ratio spectra method.
Simultaneous determination of methocarbamol and ibuprofen or diclofenac potassium using mean centering of the ratio spectra method.
Acta pharmaceutica (Zagreb, Croatia) (2012-07-04)
Nouruddin Wageih Ali, Maha Ahamd Hegazy, Mohamed Abdelkawy, Eglal Abdelahamid Abdelaleem
ABSTRACT
Accurate and sensitive methods were developed for simultaneous determination of methocarbamol and ibuprofen or diclofenac potassium in their binary mixtures, and in the presence of a methocarbamol related substance (guaifenesin) using mean centering of the ratio spectra method. The results obtained were statistically compared with the reported HPLC methods; no significant difference between the proposed methods and the reported methods was found regarding either accuracy or precision.
Sigma-Aldrich
Guaiacol glyceryl ether, ≥98% (GC)
Supelco
Guaifenesin, certified reference material, pharmaceutical secondary standard