Merck
  • ATP-dependent mitochondrial porphyrin importer ABCB6 protects against phenylhydrazine toxicity.
ATP-dependent mitochondrial porphyrin importer ABCB6 protects against phenylhydrazine toxicity.
The Journal of biological chemistry (2012-02-02)
Dagny L Ulrich, John Lynch, Yao Wang, Yu Fukuda, Deepa Nachagari, Guoqing Du, Daxi Sun, Yiping Fan, Lyudmila Tsurkan, Philip M Potter, Jerold E Rehg, John D Schuetz
ABSTRACT
Abcb6 is a mammalian mitochondrial ATP-binding cassette (ABC) transporter that regulates de novo porphyrin synthesis. In previous studies, haploinsufficient (Abcb6(+/-)) embryonic stem cells showed impaired porphyrin synthesis. Unexpectedly, Abcb6(-/-) mice derived from these stem cells appeared phenotypically normal. We hypothesized that other ATP-dependent and/or -independent mechanisms conserve porphyrins. Here, we demonstrate that Abcb6(-/-) mice lack mitochondrial ATP-driven import of coproporphyrin III. Gene expression analysis revealed that loss of Abcb6 results in up-regulation of compensatory porphyrin and iron pathways, associated with elevated protoporphyrin IX (PPIX). Phenylhydrazine-induced stress caused higher mortality in Abcb6(-/-) mice, possibly because of sustained elevation of PPIX and an inability to convert PPIX to heme despite elevated ferrochelatase levels. Therefore, Abcb6 is the sole ATP-dependent porphyrin importer, and loss of Abcb6 produces up-regulation of heme and iron pathways necessary for normal development. However, under extreme demand for porphyrins (e.g. phenylhydrazine stress), these adaptations appear inadequate, which suggests that under these conditions Abcb6 is important for optimal survival.
Sigma-Aldrich
Phenylhydrazine, 97%
Sigma-Aldrich
Phenylhydrazine hydrochloride, ≥99%