Merck
  • Homochirality beyond grinding: deracemizing chiral crystals by temperature gradient under boiling.
Homochirality beyond grinding: deracemizing chiral crystals by temperature gradient under boiling.
Chemical communications (Cambridge, England) (2011-11-05)
Cristóbal Viedma, Pedro Cintas
ABSTRACT
A single-chirality solid phase can be obtained in boiling solutions containing a racemic mixture of left- and right-handed enantiomorphous crystals due to dissolution-crystallization cycles induced by a temperature gradient. This phenomenon provides further insights into asymmetric amplification mechanisms under presumably prebiotic conditions.
Sigma-Aldrich
Sodium chlorate, ACS reagent, ≥99.0%