Merck
The thermal and enzymatic taxifolin-alphitonin rearrangement.
Journal of natural products (2011-10-14)
Paul W Elsinghorst, Taner Cavlar, Anna Müller, Annett Braune, Michael Blaut, Michael Gütschow
ABSTRACT
This report describes a detailed investigation of the thermal and enzymatic conversion of taxifolin to alphitonin. Chromatographic separation of the four dihydroquercetin stereoisomers 1-4 in combination with circular dichroism spectroscopy permitted elucidation of the kinetics of this rearrangement and characterization of the different reaction pathways involved. Our findings are corroborated by quantum chemistry calculations that reveal a unique cascade of tautomerization processes leading from taxifolin to alphitonin and also explain the racemization of alphitonin at room temperature. Furthermore, the substrate specificity toward (+)-taxifolin of an enzyme from Eubacterium ramulus catalyzing this intriguing rearrangement is demonstrated.
Product Description
Supelco
Taxifolin, analytical standard