Merck
  • Utility of routine laboratory testing in management of chronic urticaria/angioedema.
Utility of routine laboratory testing in management of chronic urticaria/angioedema.
Annals of allergy, asthma & immunology : official publication of the American College of Allergy, Asthma, & Immunology (2011-08-31)
James A Tarbox, Ravi C Gutta, Cristine Radojicic, David M Lang
ABSTRACT
Laboratory tests are routinely ordered to identify or rule out a cause in patients with chronic urticaria/angioedema (CUA). The results of these tests are usually within normal limits or unremarkable. To investigate the proportion of abnormal test results in patients with CUA leading to a change in management and in outcomes of care. Retrospective analysis of a random sample of adult patients with CUA from 2001-2009. Cases totaled 356: 166 with urticaria and angioedema (AE), 187 with urticaria, and 3 with only AE. Patients were predominately women (69.1%) and white (75.6%), with a mean age of 48 ± 15 years. Abnormalities were commonly seen in complete blood counts (34%) and in complete metabolic panels (9.4%). Among the 1,872 tests that were ordered, results of 319 (17%) were abnormal. Of 356 patients, 30 underwent further testing because of abnormalities in laboratory work. This represented 30 of 1,872 tests (1.60%). Only 1 patient benefited from a subsequent change in management. Laboratory testing in CUA patients referred for an Allergy and Immunology evaluation rarely lead to changes in management resulting in improved outcomes of care.
Sigma-Aldrich
Doxepin hydrochloride, ~85% E-isomer basis, ≥98% (GC), 15% Z-isomer basis, powder
Supelco
Doxepin hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Doxepin for system suitability, European Pharmacopoeia (EP) Reference Standard