Merck
  • Synthesis of the anti-HIV agent (-)-hyperolactone C by using oxonium ylide formation-rearrangement.
Synthesis of the anti-HIV agent (-)-hyperolactone C by using oxonium ylide formation-rearrangement.
Chemistry (Weinheim an der Bergstrasse, Germany) (2011-07-19)
David M Hodgson, Stanislav Man
ABSTRACT
Starting from readily available (S)-styrene oxide an asymmetric synthesis is described of the naturally occurring anti-HIV spirolactone (-)-hyperolactone C, which possesses adjacent fully substituted stereocenters. The key step involves a stereocontrolled Rh(II) -catalysed oxonium ylide formation-[2,3] sigmatropic rearrangement of an α-diazo-β-ketoester bearing allylic ether functionality. From the resulting furanone, an acid-catalysed lactonisation and dehydrogenation gives the natural product.
Sigma-Aldrich
(R)-(+)-Styrene oxide, ChiPros®, produced by BASF, ≥98%
Sigma-Aldrich
(R)-(+)-Styrene oxide, 97%, optical purity ee: 97% (GLC)
Sigma-Aldrich
Styrene oxide, 97%
Sigma-Aldrich
(S)-(−)-Styrene oxide, 98%, optical purity ee: 98% (GC)