Merck
  • Direct synthesis of nanoporous carbon nitride fibers using Al-based porous coordination polymers (Al-PCPs).
Direct synthesis of nanoporous carbon nitride fibers using Al-based porous coordination polymers (Al-PCPs).
Chemical communications (Cambridge, England) (2011-06-22)
Ming Hu, Julien Reboul, Shuhei Furukawa, Logudurai Radhakrishnan, Yuanjian Zhang, Pavuluri Srinivasu, Hideo Iwai, Hongjing Wang, Yoshihiro Nemoto, Norihiro Suzuki, Susumu Kitagawa, Yusuke Yamauchi
ABSTRACT
We report a new synthetic route for preparation of nanoporous carbon nitride fibers with graphitic carbon nitride polymers, by calcination of Al-based porous coordination polymers (Al-PCPs) with dicyandiamide (DCDA) under a nitrogen atmosphere.
Sigma-Aldrich
Dicyandiamide, 99%