Merck
  • Synthesis of 2-mercaptobenzothiazoles via DBU-promoted tandem reaction of o-haloanilines and carbon disulfide.
Synthesis of 2-mercaptobenzothiazoles via DBU-promoted tandem reaction of o-haloanilines and carbon disulfide.
Organic letters (2011-05-20)
Fei Wang, Shangjun Cai, Zhipeng Wang, Chanjuan Xi
ABSTRACT
An efficient strategy for the synthesis of a variety of 2-mercaptobenzothiazole derivatives has been developed. The reaction proceeded from o-haloaniline derivatives and carbon disulfide via a tandem reaction in the presence of 1,8-diazabicyclo[5.4.0]undec-7-ene (DBU) to afford the corresponding 2-mercaptobenzothiazole derivatives in good to excellent yields.
Sigma-Aldrich
2-Mercaptobenzothiazole, 97%