Merck
  • Involvement of MAPK and PI3K signaling pathway in sterigmatocystin-induced G2 phase arrest in human gastric epithelium cells.
Involvement of MAPK and PI3K signaling pathway in sterigmatocystin-induced G2 phase arrest in human gastric epithelium cells.
Molecular nutrition & food research (2011-02-03)
Xin Xing, Juan Wang, Ling Xiao Xing, Yue Hong Li, Xia Yan, Xiang Hong Zhang
ABSTRACT
Sterigmatocystin (ST), a mycotoxin commonly found in foodstuff and feedstuff, has been shown to be a carcinogenic mycotoxin in animal models. Many studies showed that the high level of ST contamination in grains might be related to the high incidence of gastric carcinoma in rural areas of China. However, up to now, the potential effects of ST on human gastric epithelium cells remain largely unknown. In this study, we explored the effects of ST on cell-cycle distribution and the regulatory mechanism in immortalized human gastric epithelium cells (GES-1). The effects of ST on the cell cycle distribution of GES-1 cells were determined with flow cytometric (FCM) analysis, Giemsa staining and immunofluorescence staining, while that on the expression of related gene-Cdc25C, Cdc2, CyclinB1 and the complex of CyclinB1-Cdc2 were studied with Western blot, reverse transcription polymerase chain reaction (RT-PCR) and immunoprecipitation assay respectively. We found that ST induced GES-1 cells arrested at G2 phase by regulating the expression of Cdc25C, Cdc2, CyclinB1 and the formation of CyclinB1-Cdc2 complex. Further study suggested JNK, ERK and PI3K/AKT/mTOR pathways to be involved in the process of G2 arrest induced by ST. The specific inhibitors of JNK and ERK reversed the role of ST, whereas that of PI3K/AKT/mTOR reinforced the effect of ST on cell-cycle distribution. This study demonstrates that JNK, ERK and PI3K/AKT/mTOR pathways participated in the G2 arrest induced by ST through the deregulation of CyclinB1, Cdc2 and Cdc25C. It may play some roles in the gastric carcinogenesis in ST exposure populations.
Product Description
Supelco
Sterigmatocystin
Sigma-Aldrich
Sterigmatocystin, powder, ≥98% (TLC)
Supelco
Sterigmatocystin solution, ~50 μg/mL in acetonitrile, analytical standard