Merck
  • Size- and shape-controlled synthesis of ZnSe nanocrystals using SeO2 as selenium precursor.
Size- and shape-controlled synthesis of ZnSe nanocrystals using SeO2 as selenium precursor.
Dalton transactions (Cambridge, England : 2003) (2010-10-27)
Huaibin Shen, Jin Zhong Niu, Hongzhe Wang, Xiaomin Li, Lin Song Li, Xia Chen
ABSTRACT
Here we report a low-cost and "green" phosphine-free route for the size- and shape-controlled synthesis of high-quality zinc blende (cubic) ZnSe nanocrystals. To avoid the use of expensive and toxic solvents such as trioctylphosphine (TOP) or tributylphosphine (TBP), SeO(2) was dispersed in 1-octadecene (ODE) as a chalcogen precursor. It has been found that the temperature and the surface ligand influenced the nucleation, the reaction speed and the formation of different shapes. Absorption spectroscopy, fluorescence spectroscopy, powder X-ray diffraction (XRD) and transmission electron microscopy (TEM) were used for the characterization of the as-synthesized ZnSe nanocrystals. The size-dependent photoluminescence (PL) range of the as-prepared ZnSe nanocrystals was between 390 and 450 nm, with the PL full width at half-maximum (FWHM) well controlled between 14 and 18 nm and PL quantum yields reached up to 40% at room temperature. Moreover, this new selenium precursor can be used to form tetrapod-shaped ZnSe nanocrystals when zinc acetylacetonate was introduced as the zinc precursor with a one-pot method.
Sigma-Aldrich
Selenium dioxide, ReagentPlus®, powder, 99.8% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Selenium dioxide, 99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Selenium dioxide, ≥99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Selenium dioxide, reagent grade, powder, 98%