Merck
  • Synthesis of C-furanosides from a D-glucal-derived cyclopropane through a ring-expansion/ring-contraction sequence.
Synthesis of C-furanosides from a D-glucal-derived cyclopropane through a ring-expansion/ring-contraction sequence.
Chemical communications (Cambridge, England) (2010-09-21)
Russell J Hewitt, Joanne E Harvey
ABSTRACT
gem-Dibromocyclopropane 1, prepared from tri-O-benzyl-D-glucal, undergoes thermal and silver-promoted ring expansion in the presence of alcohols to give substituted oxepines. With further heating, ring contraction to highly substituted tetrahydrofurans follows. These represent C-furanosides, potentially useful as precursors to C-nucleosides and other carbohydrate mimics.
Sigma-Aldrich
D-Glucal, 96%