Merck
[Studies on the chemical constituents of Solanum nigrum].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2010-09-18)
Ying Zhao, Fei Liu, Hong-Xiang Lou
ABSTRACT
To study the chemical constituents from Solanum nigrum. Compounds were isolated and purified by silica gel, Sephadex LH-20 and preparative HPLC. Their structures were identified by physicochemical properties and spectral analysis. Six compounds were isolated and identified as (+)-pinoresinol (I), (+)-syringaresinol (II), (+)-medioresinol (III), scopoletin (IV), tetracosanoic acid (V) and beta-sitosterol (VI). Compounds I - III are isolated from this genus for the first time, while compounds IV and V are isolated from this plant for the first time.
Sigma-Aldrich
Lignoceric acid, ≥99% (GC)
Sigma-Aldrich
Pinoresinol, ≥95.0% (HPLC)
Supelco
Pinoresinol, analytical standard