Merck
Cardiomyogenesis in the adult human heart.
Circulation research (2010-06-05)
Jan Kajstura, Konrad Urbanek, Shira Perl, Toru Hosoda, Hanqiao Zheng, Barbara Ogórek, João Ferreira-Martins, Polina Goichberg, Carlos Rondon-Clavo, Fumihiro Sanada, Domenico D'Amario, Marcello Rota, Federica Del Monte, Donald Orlic, John Tisdale, Annarosa Leri, Piero Anversa
ABSTRACT
The ability of the human heart to regenerate large quantities of myocytes remains controversial, and the extent of myocyte renewal claimed by different laboratories varies from none to nearly 20% per year. To address this issue, we examined the percentage of myocytes, endothelial cells, and fibroblasts labeled by iododeoxyuridine in postmortem samples obtained from cancer patients who received the thymidine analog for therapeutic purposes. Additionally, the potential contribution of DNA repair, polyploidy, and cell fusion to the measurement of myocyte regeneration was determined. The fraction of myocytes labeled by iododeoxyuridine ranged from 2.5% to 46%, and similar values were found in fibroblasts and endothelial cells. An average 22%, 20%, and 13% new myocytes, fibroblasts, and endothelial cells were generated per year, suggesting that the lifespan of these cells was approximately 4.5, 5, and 8 years, respectively. The newly formed cardiac cells showed a fully differentiated adult phenotype and did not express the senescence-associated protein p16(INK4a). Moreover, measurements by confocal microscopy and flow cytometry documented that the human heart is composed predominantly of myocytes with 2n diploid DNA content and that tetraploid and octaploid nuclei constitute only a small fraction of the parenchymal cell pool. Importantly, DNA repair, ploidy formation, and cell fusion were not implicated in the assessment of myocyte regeneration. Our findings indicate that the human heart has a significant growth reserve and replaces its myocyte and nonmyocyte compartment several times during the course of life.
Sigma-Aldrich
5-Iodo-2′-deoxyuridine, ≥99% (HPLC)