Merck
  • Enantioselective synthesis of alpha-quaternary amino acid derivatives by sequential enzymatic desymmetrization and Curtius rearrangement of alpha,alpha-disubstituted malonate diesters.
Enantioselective synthesis of alpha-quaternary amino acid derivatives by sequential enzymatic desymmetrization and Curtius rearrangement of alpha,alpha-disubstituted malonate diesters.
The Journal of organic chemistry (2010-02-06)
Violeta Iosub, Anton R Haberl, Jennifer Leung, Michael Tang, Kannan Vembaiyan, Masood Parvez, Thomas G Back
ABSTRACT
A convenient and versatile enantioselective synthesis of biologically important alpha-quaternary amino acid derivatives was based on the sequential double alkylation or arylation of dimethyl malonate, followed by desymmetrization with porcine liver esterase (PLE) and Curtius rearrangement. The PLE-mediated hydrolysis of the prochiral dialkylated malonate diesters produced the corresponding chiral half-esters in high yield and with enantiomeric excesses of 43% to >98%. Curtius rearrangement of the latter products, after trapping of the intermediate isocyanates with benzyl alcohol or amines, afforded the corresponding Cbz-protected amino esters or ureas. The absolute configurations of the major products in five examples were established by conversion to compounds with known specific rotations, or by X-ray crystallography of derivatives obtained with chiral amines of known configuration.
Sigma-Aldrich
Dimethyl malonate, purum, ≥96.0% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl malonate, 98%