Merck
  • Synthesis and characterization of N,N'-carbonyl difatty amides from palm oil.
Synthesis and characterization of N,N'-carbonyl difatty amides from palm oil.
Journal of oleo science (2009-08-06)
Emad A Jaffar Al-Mulla, Wan Md Zin Wan Yunus, Nor Azowa Bt Ibrahim, Mohd Zaki A Rahman
ABSTRACT
N,N'-Carbonyl difatty amides (CDFAs) have been synthesized from palm oil using sodium ethoxide as catalyst. Ethyl fatty esters (EFEs) were produced as a by-product as well as glycerol. The synthesis was carried out by reflux palm oil and urea in presence of ethanol. In this process, palm oil gave 79% pure CDFAs after 8 hours and molar ratio of urea to palm oil was 6.2: 1 at 78 degrees C. Both CDFAs and EFEs have been characterized using elemental analysis, Fourier transform infrared (FTIR) spectroscopy and (1)H nuclear magnetic resonance (NMR) technique.
Sigma-Aldrich
Sodium ethoxide, 95%
Sigma-Aldrich
Sodium ethoxide, technical, ≥95% (T)