Merck
  • Silver carbonate nanoparticles stabilised over alumina nanoneedles exhibiting potent antibacterial properties.
Silver carbonate nanoparticles stabilised over alumina nanoneedles exhibiting potent antibacterial properties.
Chemical communications (Cambridge, England) (2008-09-02)
Joanna J Buckley, Pratibha L Gai, Adam F Lee, Luca Olivi, Karen Wilson
ABSTRACT
A simple method of preparing Ag2CO3 nanoparticles utilising high area gamma-alumina nanoneedles has been developed; these are promising antimicrobial agents against diverse bacterial strains.
Sigma-Aldrich
Silver carbonate, 99%
Sigma-Aldrich
Silver carbonate, purum p.a., ≥99.0% (AT)
Sigma-Aldrich
Silver carbonate on Celite®, extent of labeling: ~50 wt. % loading