Merck
An application of borane as a protecting group for pyridine.
The Journal of organic chemistry (2008-08-06)
Matthew A Zajac
ABSTRACT
Efforts to couple 4 with 12 employing base mediation are problematic due to the formation of 6. To circumvent this issue, 12 was converted to the pyridine borane complex (13). Alkylation of 4 with 13 provided 3 after removal of the borane under acidic conditions and saponification of the ester.
Sigma-Aldrich
Borane pyridine complex, ~8 M BH3