Merck
  • Preparation of a DNA aptamer-Pt complex and its use in the colorimetric sensing of thrombin and anti-thrombin antibodies.
Preparation of a DNA aptamer-Pt complex and its use in the colorimetric sensing of thrombin and anti-thrombin antibodies.
Analytical chemistry (2008-07-31)
Akon Higuchi, Yi-Di Siao, Siou-Ting Yang, Pei-Vin Hsieh, Hisashi Fukushima, Yung Chang, Ruoh-Chyu Ruaan, Wen-Yih Chen
ABSTRACT
DNA aptamers carrying Pt nanoparticles were prepared by the reaction of DNA aptamers (without functionalization with biotin, thiol, or other reactive groups) with K 2[PtCl 4] in solution at 60-90 degrees C. The DNA-Pt complexes possessed peroxidase enzymatic activity while retaining the specific binding ability of the aptamers. The enzymatic reaction of these complexes obeyed Michaelis-Menten kinetics. K M for the DNA-Pt complex was found to be on the same order as K M for hemin and hemin-DNA complex but 1 or 2 orders of magnitude higher than that of horseradish peroxidase. The rate of the reaction catalyzed by the DNA-Pt complex, k cat, was found to be on the same order as that of hemin and hemin-DNA complex but 2 or 3 orders of magnitude lower than that of horseradish peroxidase. Two types of DNAzyme-linked aptamer assays (DLAAs) were developed using these complexes, which successfully detected target proteins, with the sandwich type of DLAA targeting thrombin and the competitive type of DLAA targeting anti-thrombin IgA/G/M in serum. The DNA-Pt complexes retained their peroxidase enzymatic activity even after heat treatment. DLAAs having high thermal stability were developed using these complexes, which were free of animal and plant matter because neither antibodies nor horseradish peroxidase were used in their synthesis.
Sigma-Aldrich
Potassium tetrachloroplatinate(II), ≥99.9% trace metals basis
Sigma-Aldrich
Potassium tetrachloroplatinate(II), 99.99% trace metals basis
Sigma-Aldrich
Potassium tetrachloroplatinate(II), 98%
Sigma-Aldrich
Ammonium tetrachloroplatinate(II), 99%