Merck
  • CeCl3.7H2O: an effective additive in Ru-catalyzed enantioselective hydrogenation of aromatic alpha-ketoesters.
CeCl3.7H2O: an effective additive in Ru-catalyzed enantioselective hydrogenation of aromatic alpha-ketoesters.
The Journal of organic chemistry (2008-04-24)
Qinghua Meng, Yanhui Sun, Virginie Ratovelomanana-Vidal, Jean Pierre Genêt, Zhaoguo Zhang
ABSTRACT
In the presence of catalytic amounts of CeCl 3.7H 2O, [RuCl(benzene)(S)-SunPhos]Cl is a highly effective catalyst for the asymmetric hydrogenation of aromatic alpha-ketoesters. A variety of ethyl alpha-hydroxy-alpha-arylacetates have been prepared in up to 98.3% ee with a TON up to 10,000. Challenging aromatic alpha-ketoesters with ortho substituents are also hydrogenated with high enantioselectivities. The addition of CeCl 3.7H 2O not only improves the enantioselectivity but also enhances the stability of the catalyst. The ratio of CeCl 3.7H 2O to [RuCl(benzene)(S)-SunPhos]Cl plays an important role in the hydrogenation reaction with a large substrate/catalyst ratio.
Sigma-Aldrich
Cerium(III) chloride heptahydrate, 99.9% trace metals basis
Sigma-Aldrich
Cerium(III) chloride heptahydrate, 99.999% trace metals basis
Sigma-Aldrich
Cerium(III) chloride heptahydrate, purum p.a., ≥98.0% (AT)
Sigma-Aldrich
Cerium(III) chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Cerium(III) chloride, AnhydroBeads, −10 mesh, 99.9%