- Palladium-catalyzed cyclization/Heck- and cyclization/conjugate-addition-type sequences in the preparation of polysubstituted furans.
Palladium-catalyzed cyclization/Heck- and cyclization/conjugate-addition-type sequences in the preparation of polysubstituted furans.
The Journal of organic chemistry (2008-03-28)
José M Aurrecoechea, Aritz Durana, Elena Pérez
PMID18363372
ABSTRACT
Palladium-catalyzed heterocyclization-coupling sequences have been developed starting from buta-1,2,3-trienyl carbinols and electron-deficient alkenes. Polysubstituted furans are formed where the heterocyclic ring originates from the elements of the butatrienyl carbinol while the electron-deficient olefin is incorporated as a C-3 substituent. In most cases, the reaction proceeds via a Heck-type pathway leading to the efficient formation of 3-vinylfurans. However, couplings with methyl vinyl ketone display a divergent behavior to afford selectively either Heck- or hydroarylation-type products depending on reaction conditions.
Sigma-Aldrich
3-Buten-2-one, contains 0.3-1.0% hydroquinone as stabilizer, technical grade, 90%