Merck
  • [Method study on dehydroaetic acid in food by high performance liquid chromatography].
[Method study on dehydroaetic acid in food by high performance liquid chromatography].
Wei sheng yan jiu = Journal of hygiene research (2008-02-29)
Huizhen Zhang, Aiguo Ma, Hui Liang, Yongye Sun
ABSTRACT
To estabish a high performance liquid chromatographic (HPLC) method for the determination of dehydroaetic acid in food determinated. Separation was carried out on a column of C18 (51 microm, 250 x 4.60mm) at the temperature of 25 degrees C with methanol/0.02mol/L ammonium acetate (5:95, V/V) as mobile phase, and elution flow rate was 1.0ml/min. The detection wavelength was 306nm and the injection volume was 10 microl, quantitation was used by external standard method. The results showed a good linear relationship between the peak area and the concentration (r = 0.9997), the recoveries for Dehydroaetic acid products were 96.5% - 99.5%, and the relative standard deviation was 1.09% - 5.85% for different kinds of food. The results showed that the method may be simple, rapid, accurate and applicable for the determination of dehydroaetic acid in food.
Sigma-Aldrich
Dehydroacetic acid, ≥98.0% (T)