Merck
  • Liquid chromatographic direct resolution of beta-amino acids on a doubly tethered chiral stationary phase containing N--H amide linkage based on (+)-(18-crown-6)- 2,3,11,12-tetracarboxylic acid.
Liquid chromatographic direct resolution of beta-amino acids on a doubly tethered chiral stationary phase containing N--H amide linkage based on (+)-(18-crown-6)- 2,3,11,12-tetracarboxylic acid.
Chirality (2007-07-07)
Myung Ho Hyun, Yanci Song, Yoon Jae Cho, Hee Jung Choi
ABSTRACT
A doubly tethered chiral stationary phase (CSP) prepared by bonding (+)-(18-crown-6)-2,3,11,12-tetracarboxylic acid to doubly tethered primary aminoalkyl silica gel was used for the resolution of various beta-amino acids. All the beta-amino acids tested were resolved quite well, the separation (alpha) and the resolution factors (RS) being in the ranges 1.34-2.09 and 2.52-7.45, respectively, with a mobile phase of methanol-water (50:50, v/v) containing 10 mM acetic acid. The chiral recognition efficiency of the doubly-tethered CSP was found to be generally superior to that of the corresponding singly-tethered CSP in the resolution of beta-amino acids. The chiral recognition behaviors for the resolution of beta-amino acids on the doubly tethered CSP were examined by varying the type and content of organic and acidic modifiers in the aqueous mobile phase and the column temperature.