- Iron porphyrins catalyze the synthesis of non-protected amino acid esters from ammonia and diazoacetates.
Iron porphyrins catalyze the synthesis of non-protected amino acid esters from ammonia and diazoacetates.
Chemical communications (Cambridge, England) (2007-02-08)
Iris Aviv, Zeev Gross
PMID17283790
ABSTRACT
Iron complexes of porphyrins (and corroles to a lesser extent) are the first catalysts to utilize ammonia for the synthesis of N-free amino acid esters.