Merck
  • Hydroaminomethylation with novel rhodium-carbene complexes: an efficient catalytic approach to pharmaceuticals.
Hydroaminomethylation with novel rhodium-carbene complexes: an efficient catalytic approach to pharmaceuticals.
Chemistry (Weinheim an der Bergstrasse, Germany) (2006-11-09)
Moballigh Ahmed, Cathleen Buch, Lucie Routaboul, Ralf Jackstell, Holger Klein, Anke Spannenberg, Matthias Beller
ABSTRACT
Starting from [{Rh(cod)Cl}(2)] and 1,3-dimesitylimidazole-2-ylidenes the novel [RhCl(cod)(carbene)] complexes 1-5 have been synthesized, characterized, and tested in the hydroaminomethylation of aromatic olefins. The influence of different ligands and reaction parameters on the catalytic activity was investigated in detail applying 1,1-diphenylethylene and piperidine as a model system. The scope and limitations of the novel catalysts is shown in the preparation of 16 biologically active 1-amino-3,3-diarylpropenes. In general, high chemo- and regioselectivity as well as good yields of the desired products were achieved.
Sigma-Aldrich
1,1-Diphenylethylene, 97%