Merck
  • Cationic long-chain ceramide LCL-30 induces cell death by mitochondrial targeting in SW403 cells.
Cationic long-chain ceramide LCL-30 induces cell death by mitochondrial targeting in SW403 cells.
Molecular cancer therapeutics (2006-07-05)
Daniel Dindo, Felix Dahm, Zdzislaw Szulc, Alicja Bielawska, Lina M Obeid, Yusuf A Hannun, Rolf Graf, Pierre-Alain Clavien
ABSTRACT
Ceramides are sphingolipid second messengers that are involved in the mediation of cell death. There is accumulating evidence that mitochondria play a central role in ceramide-derived toxicity. We designed a novel cationic long-chain ceramide [omega-pyridinium bromide D-erythro-C16-ceramide (LCL-30)] targeting negatively charged mitochondria. Our results show that LCL-30 is highly cytotoxic to SW403 cells (and other cancer cell lines) and preferentially accumulates in mitochondria, resulting in a decrease of the mitochondrial membrane potential, release of mitochondrial cytochrome c, and activation of caspase-3 and caspase-9. Ultrastructural analyses support the concept of mitochondrial selectivity. Interestingly, levels of endogenous mitochondrial C16-ceramide decreased by more than half, whereas levels of sphingosine-1-phosphate increased dramatically and selectively in mitochondria after administration of LCL-30, suggesting the presence of a mitochondrial sphingosine kinase. Of note, intracellular long-chain ceramide levels and sphingosine-1-phosphate remained unaffected in the cytosolic and extramitochondrial (nuclei/cellular membranes) cellular fractions. Furthermore, a synergistic effect of cotreatment of LCL-30 and doxorubicin was observed, which was not related to alterations in endogenous ceramide levels. Cationic long-chain pyridinium ceramides might be promising new drugs for cancer therapy through their mitochondrial preference.
Sigma-Aldrich
N-Palmitoyl-D-sphingosine, ≥98.0% (TLC)