Merck
  • Epidermal growth factor-conjugated poly(ethylene glycol)-block- poly(delta-valerolactone) copolymer micelles for targeted delivery of chemotherapeutics.
Epidermal growth factor-conjugated poly(ethylene glycol)-block- poly(delta-valerolactone) copolymer micelles for targeted delivery of chemotherapeutics.
Bioconjugate chemistry (2006-03-16)
Faquan Zeng, Helen Lee, Christine Allen
ABSTRACT
Epidermal growth factor (EGF)-conjugated copolymer micelles were prepared from a mixture of diblock copolymers of methoxy poly(ethylene glycol)-block-poly(delta-valerolactone) (MePEG-b-PVL) and EGF-PEG-b-PVL for targeted delivery to EGF receptor (EGFR)-overexpressing cancers. The block copolymers and functionalized block copolymers were synthesized using PEG as the macroinitiator and HCl-diethyl ether as the catalyst. The MePEG-b-PVL and the carboxyl-terminated PEG-b-PVL (HOOC-PEG-b-PVL) copolymers were found to have molecular weights of 5940 and 5900, respectively, as determined by gel permeation chromatography (GPC) analyses. The HOOC-PEG-b-PVL copolymers were then activated by N-hydroxysuccinimide and subsequently reacted with EGF to form the EGF-PEG-b-PVL copolymers. The efficiency for the conjugation of EGF to the copolymer was found to be 60.9%. A hydrophobic fluorescent probe, CM-DiI, was loaded into both the nontargeted MePEG-b-PVL micelles and the targeted EGF-conjugated PEG-b-PVL micelles. The effective mean diameters of the CMDiI-loaded nontargeted and the CMDiI-loaded targeted micelles were found to be 32 +/- 1 nm and 45 +/- 2 nm, respectively, as determined by dynamic light scattering (DLS). The zeta potentials for the nontargeted micelles (no CM-DiI-loaded), CM-DiI-loaded nontargeted micelles, and CM-DiI-loaded targeted micelles were found to be -6.5, -8.7, and - 13.5 mV, respectively. Evaluation of the in vitro release of CM-DiI from the MePEG-b-PVL micelles in phosphate buffer saline (0.01 M, pH = 7.4) containing 10% (v/v) fetal bovine serum at 37 degrees C revealed that approximately 20% of the probe was released within the first 2 h. Confocal laser scanning microscopy (CLSM) analysis revealed that the targeted micelles containing CM-DiI accumulated intracellularly in EGFR-overexpressing MDA-MB-468 breast cancer cells following a 2 h incubation period, while no detectable cell uptake was observed for the nontargeted micelles. Results obtained from the confocal images were confirmed in an independent study by measuring the intracellular CM-DiI fluorescence in cell lysate. In addition, the presence of free EGF was found to decrease the extent of uptake of the targeted micelles. Nuclear staining of the cells with Hoechst 33258 indicated that the targeted micelles mainly localized in the perinuclear region and some of the micelles were localized in the nucleus. These results demonstrate that the EGF-conjugated copolymer micelles developed in this study have potential as vehicles for targeting hydrophobic drugs to EGFR-overexpressing cancers.
Sigma-Aldrich
δ-Valerolactone, technical grade