Merck
  • Sensitive determination of 4-(4-chlorophenyl)-4-hydroxypiperidine, a metabolite of haloperidol, in a rat biological sample by HPLC with fluorescence detection after pre-column derivatization using 4-fluoro-7-nitro-2,1,3-benzoxadiazole.
Sensitive determination of 4-(4-chlorophenyl)-4-hydroxypiperidine, a metabolite of haloperidol, in a rat biological sample by HPLC with fluorescence detection after pre-column derivatization using 4-fluoro-7-nitro-2,1,3-benzoxadiazole.
Biomedical chromatography : BMC (2006-03-01)
Yasuhiko Higashi, Shota Nakamura, Youichi Fujii
ABSTRACT
4-(4-Chlorophenyl)-4-hydroxypiperidine (CPHP), one of the metabolites of haloperidol, is considered to exhibit brain toxicity. CPHP concentrations in plasma and tissue homogenates (each 200 microL) from rats were analyzed by HPLC fluorescence detection after pre-column derivatization with 4-fluoro-7-nitro-2,1,3-benzoxadiazole (NBD-F). After basic extraction of the samples with benzene, the derivatization with NBD-F was conducted in borate buffer (pH 8.0) at 60 degrees C for 3 min. Mexiletine was carried through the procedure as an internal standard. The regression equation for CPHP showed a good linearity in the range of 0.03-1 microg/mL with a detection limit of 0.008 microg/mL. The coefficient of variation was less than 11.6%. Plasma concentration-time courses of CPHP after intraperitoneal or per oral administration of CPHP, haloperidol or reduced haloperidol were examined, and the pharmacokinetic parameters were estimated. Additionally, CPHP levels in various tissues at 8 h after intraperitoneal administration of these compounds were compared. The method was simple and sensitive, useful for determination of CPHP in rat biological samples using as little as 200 microL of sample volume and could be applied for pharmacokinetic study.
Sigma-Aldrich
4-(4-Chlorophenyl)-4-hydroxypiperidine, 99%